Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013856.1 | NP_001013878.1 | 13 | 1880 | ||
GenBank mRNA | BC079308.1 | - | 13 | 1880 | 7134677 |