Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193307.1 | NP_001180236.1 | 2 | 3364 | ||
NM_017654.3 | NP_060124.2 | 3 | 3465 | |||
GenBank mRNA | AB095925.2 | - | 3 | 3399 | ||
AF445355.1 | - | 3 | 3465 | |||
AK000080.1 | - | 1 | 334 | |||
AK122951.1 | - | 2 | 3302 | |||
AK125101.1 | - | 1 | 824 | |||
AK125131.1 | - | 1 | 1514 | |||
AK225201.1 | - | 1 | 334 | |||
BC132773.1 | - | 2 | 3295 | 40146795 | ||
BC132775.1 | - | 2 | 3295 | 40146797 | ||
BC150249.1 | - | 3 | 3420 | 9007474 | ||
BX647072.1 | - | 3 | 3421 |