Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013867.1 | NP_001013889.1 | 10 | 1357 | ||
XM_006248858.2 | XP_006248920.1 | 11 | 2184 | |||
XM_006248859.2 | XP_006248921.1 | 11 | 2052 | |||
XM_006248860.2 | XP_006248922.1 | 11 | 2184 | |||
XM_006248861.2 | XP_006248923.1 | 10 | 2093 | |||
GenBank mRNA | BC081935.1 | - | 10 | 1357 | 7114952 | |
FQ212099.1 | - | 10 | 1464 |