Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Desi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013873.1 | NP_001013895.1 | 5 | 2028 | ||
XM_006250316.2 | XP_006250378.1 | 5 | 2378 | |||
XM_006250317.2 | XP_006250379.1 | 6 | 2792 | |||
XM_008769783.1 | XP_008768005.1 | 5 | 2335 | |||
XM_008769784.1 | XP_008768006.1 | 6 | 2511 | |||
GenBank mRNA | BC083584.1 | - | 5 | 2028 | 7131107 |