Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam149b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013878.1 | NP_001013900.1 | 13 | 2037 | ||
XM_006251632.2 | XP_006251694.1 | 14 | 2308 | |||
XM_006251634.2 | XP_006251696.1 | 12 | 1938 | |||
GenBank mRNA | BC087126.1 | - | 13 | 2037 | 7111155 |