Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300946.1 | NP_001287875.1 | 11 | 2059 | ||
NM_017660.3 | NP_060130.3 | 12 | 2362 | |||
XM_011528105.1 | XP_011526407.1 | 12 | 2248 | |||
XM_017026901.1 | XP_016882390.1 | 11 | 2137 | |||
XM_017026902.1 | XP_016882391.1 | 13 | 2311 | |||
XM_017026903.1 | XP_016882392.1 | 13 | 2401 | |||
XM_017026904.1 | XP_016882393.1 | 13 | 2358 | |||
XM_017026905.1 | XP_016882394.1 | 13 | 2371 | |||
XM_017026906.1 | XP_016882395.1 | 11 | 2173 | |||
XM_017026907.1 | XP_016882396.1 | 14 | 2480 | |||
XM_017026908.1 | XP_016882397.1 | 15 | 2740 | |||
XM_017026909.1 | XP_016882398.1 | 15 | 2691 | |||
XM_017026910.1 | XP_016882399.1 | 13 | 2300 | |||
XM_017026911.1 | XP_016882400.1 | 13 | 2472 | |||
GenBank mRNA | AK000092.1 | - | 4 | 804 | ||
AK125974.1 | - | 14 | 2470 | |||
AK225198.1 | - | 4 | 804 | |||
AK296781.1 | - | 8 | 1620 | |||
AK303482.1 | - | 7 | 1570 | |||
AL390164.1 | - | 3 | 585 | |||
BC011684.1 | - | 12 | 2091 | 3953055 | ||
BC012902.1 | - | 5 | 950 |