Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017664.3 | NP_060134.2 | 6 | 1150 | ||
NR_104587.1 | 7 | 1212 | ||||
GenBank mRNA | AK000100.1 | - | 6 | 1112 | ||
AK000111.1 | - | 1 | 443 | |||
AK026017.1 | - | 2 | 224 | |||
AK095763.1 | - | 4 | 822 | |||
AK096083.1 | - | 3 | 520 | |||
AK123341.1 | - | 1 | 985 | |||
AK125100.1 | - | 1 | 2118 | |||
BX641022.1 | - | 1 | 2146 |