Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300816.2 | NP_001287745.1 | 4, 5 | 372 | ||
NM_001300822.2 | NP_001287751.1 | 4, 5 | 400 | |||
NM_001308127.1 | NP_001295056.1 | 4, 5 | 402 | |||
NM_001308128.1 | NP_001295057.1 | 2, 3 | 144 | |||
NM_001308130.1 | NP_001295059.1 | 3, 4 | 334 | |||
NM_017665.3 | NP_060135.1 | 3, 4 | 306 | |||
NR_125337.2 | 3, 4 | 310 | ||||
NR_131766.1 | 4, 5 | 455 | ||||
NR_131767.1 | 3, 4 | 338 | ||||
XM_011543503.2 | XP_011541805.1 | 4, 5 | 447 | |||
GenBank mRNA | AF086446.1 | - | 2, 3 | 188 | ||
AK000101.1 | - | 3, 4 | 250 | |||
BC005211.1 | - | 3, 4 | 263 | 3961163 | ||
BC022443.1 | - | 4, 5 | 313 | 4285562 | ||
BF983215.1 | - | 3, 4 | 268 | |||
BG699335.1 | - | 4, 5 | 380 | |||
BI826979.1 | - | 3, 4 | 306 | |||
BM470321.1 | - | 3, 4 | 257 | |||
CB112418.1 | - | 4, 5 | 341 | |||
DA345717.1 | - | 2, 3 | 112 | |||
DA466245.1 | - | 4, 5 | 396 | |||
DA528172.1 | - | 3, 4 | 278 | |||
DA905071.1 | - | 4, 5 | 347 |