Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdilt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013902.1 | NP_001013924.1 | 2 | 304 | ||
XM_008759714.1 | XP_008757936.1 | 2 | 321 | |||
GenBank mRNA | BC083897.1 | - | 2 | 304 | 7133354 |