Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013926.2 | NP_001013948.2 | 5 | 688 | ||
XM_003749789.3 | XP_003749837.1 | 5 | 639 | |||
XM_006236804.2 | XP_006236866.1 | 6 | 1345 | |||
XM_006236805.2 | XP_006236867.1 | 6 | 840 | |||
XM_006236808.2 | XP_006236870.1 | 6 | 710 | |||
GenBank mRNA | CK475733.1 | - | 5 | 688 | ||
CV112449.1 | - | 5 | 616 | |||
FQ234910.1 | - | 5 | 645 |
Gene Symbol: LOC103690067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236742.2 | XP_006236804.1 | 1345 | |||
XM_006236743.2 | XP_006236805.1 | 840 | ||||
XM_006236745.2 | XP_006236807.1 | 710 | ||||
XM_006236746.2 | XP_006236808.1 | 624 | ||||
XM_008763108.1 | XP_008761330.1 | 724 |