Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013944.1 | NP_001013966.1 | 11 | 1314 | ||
XM_006240231.1 | XP_006240293.1 | 11 | 1403 | |||
GenBank mRNA | BC079166.1 | - | 11 | 1314 | 7129668 | |
FQ213708.1 | - | 11 | 1327 |