Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013947.1 | NP_001013969.1 | 10 | 1615 | ||
XM_008765502.1 | XP_008763724.1 | 10 | 1583 | |||
GenBank mRNA | AB559554.1 | - | 10 | 1612 | ||
BC081982.1 | - | 10 | 1615 | 7128584 |