Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013958.1 | NP_001013980.1 | 11 | 1281 | ||
XM_006256710.2 | XP_006256772.1 | 11 | 1453 | |||
XM_006256712.2 | XP_006256774.1 | 11 | 1564 | |||
XM_006256713.2 | XP_006256775.1 | 11 | 1338 | |||
XM_006256714.2 | XP_006256776.1 | 11 | 1458 | |||
XM_008773071.1 | XP_008771293.1 | 11 | 1351 | |||
GenBank mRNA | BC085816.1 | - | 11 | 1281 | 7123612 | |
FQ225212.1 | - | 6 | 945 | |||
FQ232559.1 | - | 6 | 945 | |||
FQ233394.1 | - | 6 | 945 |