Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013960.1 | NP_001013982.1 | 11 | 978 | ||
GenBank mRNA | BC085822.1 | - | 11 | 978 | 7126662 |