Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013964.1 | NP_001013986.1 | 17 | 2162 | ||
XM_006245792.2 | XP_006245854.1 | 17 | 2187 | |||
XM_006245793.2 | XP_006245855.1 | 17 | 2184 | |||
XM_006245794.2 | XP_006245856.1 | 16 | 2120 | |||
XM_006245795.2 | XP_006245857.1 | 16 | 2117 | |||
XM_006245796.2 | XP_006245858.1 | 18 | 2232 | |||
XM_008767485.1 | XP_008765707.1 | 18 | 2228 | |||
XM_008767486.1 | XP_008765708.1 | 17 | 2162 | |||
GenBank mRNA | BC083621.1 | - | 17 | 2162 | 7134018 |