Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013966.1 | NP_001013988.1 | 2 | 225 | ||
GenBank mRNA | BC087596.1 | - | 2 | 225 | 7310343 |