Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135589.2 | NP_001129061.1 | 3 | 488 | ||
NM_017686.3 | NP_060156.1 | 3 | 488 | |||
XM_005270959.2 | XP_005271016.1 | 3 | 488 | |||
XM_011541621.2 | XP_011539923.1 | 3 | 488 | |||
XM_011541622.1 | XP_011539924.1 | 3 | 376 | |||
XM_011541623.2 | XP_011539925.1 | 3 | 488 | |||
XM_011541624.2 | XP_011539926.1 | 3 | 851 | |||
XM_017001531.1 | XP_016857020.1 | 3 | 430 | |||
XM_017001532.1 | XP_016857021.1 | 3 | 851 | |||
GenBank mRNA | AK000149.1 | - | 3 | 391 | ||
BC013132.1 | - | 3 | 371 | 3448678 | ||
BJ992034.1 | - | 3 | 411 | |||
DB043931.1 | - | 3 | 491 |