Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017446.1 | NP_001017446.1 | 5 | 865 | ||
GenBank mRNA | BC079333.1 | - | 2 | 482 | ||
BC091281.1 | - | 5 | 865 | 7192011 |