Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017449.1 | NP_001017449.1 | 8 | 1034 | ||
XM_006233214.1 | XP_006233276.1 | 8 | 847 | |||
XM_008761454.1 | XP_008759676.1 | 7 | 895 | |||
GenBank mRNA | BC092598.1 | - | 8 | 1034 | 7321341 |