Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ervfrd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024239.1 | NP_001019410.1 | 1 | 610 | ||
XM_008770821.1 | XP_008769043.1 | 2 | 1422 | |||
XM_008770822.1 | XP_008769044.1 | 2 | 1016 |