Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024241.1 | NP_001019412.1 | 2 | 761 | ||
GenBank mRNA | AB196531.1 | - | 2 | 761 |