Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024645.1 | NP_001019816.1 | 23 | 2063 | ||
GenBank mRNA | AK131164.1 | - | 23 | 2006 | ||
AK148355.1 | - | 23 | 2086 | |||
AK158466.1 | - | 24 | 2309 | |||
AK195273.1 | - | 2 | 87 | |||
BC036313.1 | - | 8 | 625 | |||
BC060252.1 | - | 4 | 603 | |||
BC082805.1 | - | 23 | 1983 | |||
BC092223.1 | - | 23 | 1981 |