Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024740.1 | NP_001019911.1 | 3 | 270 | ||
XM_006248676.2 | XP_006248738.1 | 3 | 392 | |||
GenBank mRNA | BC097459.1 | - | 3 | 270 | 7457223 |