Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024746.1 | NP_001019917.1 | 10, 11 | 1046 | ||
XM_006255078.1 | XP_006255140.1 | 11, 12 | 1422 | |||
XM_008772277.1 | XP_008770499.1 | 11, 12 | 1363 | |||
XM_008772278.1 | XP_008770500.1 | 11, 12 | 1494 | |||
XM_008772279.1 | XP_008770501.1 | 11, 12 | 1545 | |||
XM_008772280.1 | XP_008770502.1 | 11, 12 | 1423 | |||
GenBank mRNA | AY953248.1 | - | 10, 11 | 1046 | ||
BC161823.1 | - | 10, 11 | 1292 |