Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024752.1 | NP_001019923.1 | 6, 7 | 744 | ||
GenBank mRNA | BC097481.1 | - | 6, 7 | 744 | 7459091 |