Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024757.1 | NP_001019928.1 | 25 | 3039 | ||
XM_006237930.2 | XP_006237992.1 | 25 | 2954 | |||
XM_006237931.2 | XP_006237993.1 | 24 | 3381 | |||
XM_008763590.1 | XP_008761812.1 | 26 | 3252 | |||
XM_008763591.1 | XP_008761813.1 | 18 | 2447 | |||
GenBank mRNA | BC097386.1 | - | 25 | 3039 | 7446512 | |
FQ211113.1 | - | 2 | 159 | |||
FQ216775.1 | - | 10 | 1130 |