Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024765.1 | NP_001019936.1 | 13, 14 | 1706 | ||
XM_006247321.2 | XP_006247383.1 | 13, 14 | 1988 | |||
XM_006247322.2 | XP_006247384.1 | 13, 14 | 1903 | |||
XM_006247323.2 | XP_006247385.1 | 13, 14 | 1760 | |||
XM_006247324.2 | XP_006247386.1 | 13, 14 | 1707 | |||
XM_006247325.2 | XP_006247387.1 | 13, 14 | 1863 | |||
XM_006247326.2 | XP_006247388.1 | 13, 14 | 1858 | |||
GenBank mRNA | BC095903.1 | - | 13, 14 | 1706 | 7096697 | |
FQ211883.1 | - | 13, 14 | 1706 |