Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024865.1 | NP_001020036.1 | 4 | 1430 | ||
GenBank mRNA | BC097472.1 | - | 4 | 1430 | 7457772 |