Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pih1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024868.1 | NP_001020039.1 | 4, 5 | 729 | ||
XM_006229027.1 | XP_006229089.1 | 5, 6 | 466 | |||
XM_006229028.2 | XP_006229090.1 | 5, 6 | 702 | |||
GenBank mRNA | BC097946.1 | - | 4, 5 | 729 | 7366310 |