Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 45, 46 | 5258 | ||
NM_014689.2 | NP_055504.2 | 45, 46 | 5401 | |||
XM_005246673.4 | XP_005246730.1 | 45, 46 | 5260 | |||
XM_006712617.3 | XP_006712680.1 | 45, 46 | 5425 | |||
XM_017004441.1 | XP_016859930.1 | 45, 46 | 5428 | |||
XM_017004442.1 | XP_016859931.1 | 45, 46 | 5404 | |||
XM_017004443.1 | XP_016859932.1 | 45, 46 | 5477 | |||
XM_017004444.1 | XP_016859933.1 | 44, 45 | 5384 | |||
XM_017004445.1 | XP_016859934.1 | 45, 46 | 5258 | |||
XM_017004446.1 | XP_016859935.1 | 44, 45 | 5282 | |||
XM_017004447.1 | XP_016859936.1 | 45, 46 | 5400 | |||
GenBank mRNA | AF250396.1 | - | 5, 6 | 770 | ||
AK000227.1 | - | 3, 4 | 370 | |||
AK293847.1 | - | 11, 12 | 1327 | |||
AK293879.1 | - | 16, 17 | 1807 | |||
AK307914.1 | - | 5, 6 | 762 | |||
BC015018.1 | - | 3, 4 | 368 | |||
CR749492.1 | - | 20, 21 | 2246 | |||
EU236710.2 | - | 45, 46 | 5255 |