Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024965.1 | NP_001020136.1 | 6 | 523 | ||
XM_006256857.1 | XP_006256919.1 | 6 | 613 |