Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024970.2 | NP_001020141.2 | 5 | 653 | ||
GenBank mRNA | BC091401.1 | - | 2 | 536 | 7324146 | |
CB724808.1 | - | 2 | 108 |