Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024982.1 | NP_001020153.1 | 5, 7 | 390 | ||
XM_006237022.2 | XP_006237084.1 | 5, 7 | 1370 | |||
GenBank mRNA | BC091347.1 | - | 5, 7 | 390 | 7316601 |