Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024984.1 | NP_001020155.1 | 8 | 689 | ||
GenBank mRNA | BC083597.1 | - | 8 | 689 | 7132841 | |
FQ213743.1 | - | 9 | 656 | |||
FQ227558.1 | - | 8 | 553 | |||
FQ229098.1 | - | 10 | 771 | |||
FQ230170.1 | - | 10 | 772 |