Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025067.1 | NP_001020238.1 | 13 | 1950 | ||
GenBank mRNA | AK028527.1 | - | 3 | 567 | ||
AK036116.1 | - | 13 | 1952 | |||
AK155089.1 | - | 13 | 2013 | |||
AK173030.1 | - | 3 | 848 | |||
BC094228.1 | - | 13 | 1887 | 30362610 | ||
BC138422.1 | - | 13 | 1861 | 8861442 |