Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017734.4 | NP_060204.1 | 3 | 583 | ||
GenBank mRNA | AF262379.1 | - | 3 | 514 | ||
AJ312214.1 | - | 3 | 425 | |||
AK000278.1 | - | 3 | 345 | |||
AL390971.1 | - | 3 | 513 | |||
AL390972.1 | - | 3 | 116 | |||
AL390973.1 | - | 3 | 309 | |||
BC012570.1 | - | 3 | 398 | 4245261 | ||
KU178657.1 | - | 3 | 140 | |||
KU178658.1 | - | 3 | 140 |