Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164086.1 | NP_001157558.1 | 9 | 1431 | ||
NM_001164087.1 | NP_001157559.1 | 9 | 1447 | |||
NM_011983.2 | NP_036113.1 | 9 | 1464 | |||
GenBank mRNA | AB017136.1 | - | 9 | 1457 | ||
AF093259.1 | - | 9 | 1336 | |||
AF093260.1 | - | 9 | 1369 | |||
AK003632.1 | - | 1 | 376 | |||
AK158673.1 | - | 1533 | ||||
BC038314.1 | - | 5 | 1105 |