Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287164.1 | NP_001274093.1 | 19 | 2639 | ||
NM_001287165.1 | NP_001274094.1 | 19 | 2588 | |||
NM_001287166.1 | NP_001274095.1 | 19 | 2588 | |||
NM_001287167.1 | NP_001274096.1 | 19 | 2619 | |||
NM_022979.2 | NP_075355.1 | 19 | 2639 | |||
GenBank mRNA | AK146491.1 | - | 19 | 2620 | ||
AK164402.1 | - | 19 | 2574 | |||
BC039912.1 | - | 4 | 574 | |||
BC050911.1 | - | 19 | 2593 | 6417631 | ||
BC057608.1 | - | 19 | 2593 | 6417631 | ||
BC078630.1 | - | 5 | 642 | |||
BC112912.1 | - | 19 | 2593 | 5697919 | ||
BC137975.1 | - | 18 | 2447 | 8860994 | ||
BC145071.1 | - | 18 | 2447 | 9053619 |