Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000309.1 | NP_001000309.1 | 1 | 571 |
Gene Symbol: LOC100909900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749507.2 | XP_003749555.1 | 1 | 571 |