Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017753.2 | NP_060223.2 | 3 | 413 | ||
NM_207299.1 | NP_997182.1 | 3 | 546 | |||
GenBank mRNA | AK000307.1 | - | 3 | 413 | ||
AK225306.1 | - | 3 | 413 | |||
AK303718.1 | - | 1 | 293 | |||
AK303807.1 | - | 2 | 409 | |||
AK309115.1 | - | 1 | 122 | |||
AY304515.1 | - | 3 | 279 | |||
AY337718.1 | - | 2 | 159 | |||
BC022465.1 | - | 3 | 279 | 4797229 | ||
BI601921.1 | - | 3 | 552 | |||
BI914341.1 | - | 3 | 586 | |||
CA772610.1 | - | 3 | 449 |