Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000802.1 | NP_001000802.1 | 1 | 622 |
Gene Symbol: LOC100910145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749185.2 | XP_003749233.2 | 1 | 640 |