Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173977.1 | NP_001167448.1 | 37 | 4927 | ||
NM_017640.5 | NP_060110.4 | 37 | 4945 | |||
XM_005249218.1 | XP_005249275.1 | 38 | 5095 | |||
XM_017011008.1 | XP_016866497.1 | 40 | 5125 | |||
XM_017011009.1 | XP_016866498.1 | 40 | 5122 | |||
XM_017011010.1 | XP_016866499.1 | 39 | 5113 | |||
XM_017011011.1 | XP_016866500.1 | 39 | 5107 | |||
XM_017011012.1 | XP_016866501.1 | 40 | 5107 | |||
XM_017011013.1 | XP_016866502.1 | 39 | 5089 | |||
XM_017011014.1 | XP_016866503.1 | 39 | 5065 | |||
XM_017011015.1 | XP_016866504.1 | 39 | 4975 | |||
XM_017011016.1 | XP_016866505.1 | 38 | 4957 | |||
GenBank mRNA | AI142012.1 | - | 1 | 291 | ||
AK000055.1 | - | 11 | 2139 | |||
BC140770.1 | - | 37 | 4701 | 9021638 | ||
CB132825.1 | - | 1 | 320 |