Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009909.3 | NP_001009909.2 | 12 | 1678 | ||
NM_001252008.1 | NP_001238937.1 | 12 | 1674 | |||
NM_001252010.1 | NP_001238939.1 | 10 | 1552 | |||
GenBank mRNA | AK126974.1 | - | 4 | 721 | ||
AL832641.3 | - | 12 | 1590 | |||
BC132829.1 | - | 12 | 1470 | 40146851 | ||
BC144177.1 | - | 12 | 1466 |