Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010851.2 | NP_001010851.1 | 4 | 716 | ||
XM_011527460.1 | XP_011525762.1 | 4 | 1043 | |||
XM_017027440.1 | XP_016882929.1 | 4 | 911 | |||
XM_017027441.1 | XP_016882930.1 | 2 | 780 | |||
GenBank mRNA | AK022545.1 | - | 1 | 286 | ||
AK024074.1 | - | 5 | 768 | |||
AK024268.1 | - | 1 | 353 | |||
AK097729.1 | - | 5 | 1097 | |||
BC136833.1 | - | 4 | 704 | 9020824 | ||
BC136834.1 | - | 4 | 704 | 9020825 | ||
BX538183.1 | - | 4 | 685 | |||
BX648899.1 | - | 4 | 717 |