Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010887.2 | NP_001010887.2 | 2, 3 | 386 | ||
XM_005251448.3 | XP_005251505.1 | 3, 4 | 437 | |||
XM_011517858.1 | XP_011516160.1 | 2, 3 | 386 | |||
XM_017014694.1 | XP_016870183.1 | 2, 3 | 142 | |||
GenBank mRNA | AF370405.1 | - | 3, 4 | 298 | ||
AK123581.1 | - | 2, 3 | 386 | |||
AY312516.1 | - | 2, 3 | 228 | |||
BC092487.1 | - | 3, 4 | 437 | 30528463 |