Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010888.3 | NP_001010888.3 | 5 | 4184 | ||
XM_011530939.2 | XP_011529241.1 | 6 | 4429 | |||
XM_017029478.1 | XP_016884967.1 | 11 | 5695 | |||
XM_017029479.1 | XP_016884968.1 | 13 | 8988 | |||
XM_017029480.1 | XP_016884969.1 | 11 | 5729 | |||
XM_017029481.1 | XP_016884970.1 | 11 | 5724 | |||
XM_017029482.1 | XP_016884971.1 | 10 | 5634 | |||
XM_017029483.1 | XP_016884972.1 | 9 | 5430 | |||
XM_017029484.1 | XP_016884973.1 | 9 | 5062 | |||
GenBank mRNA | BX647241.1 | - | 7 | 3934 | ||
DA401831.1 | - | 1 | 481 |