Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012134.1 | NP_001012134.1 | 545 | |||
GenBank mRNA | BC081994.1 | - | 545 | 7130315 |