Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012136.1 | NP_001012136.1 | 9 | 1462 | ||
XM_006239740.2 | XP_006239802.1 | 10 | 2152 | |||
XM_006239741.2 | XP_006239803.1 | 9 | 2051 | |||
XM_008764478.1 | XP_008762700.1 | 12 | 2015 | |||
GenBank mRNA | BC082104.1 | - | 9 | 1462 | 7134703 |