Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012418.3 | NP_001012418.2 | 13 | 2019 | ||
XM_005249078.3 | XP_005249135.2 | 13 | 1984 | |||
XM_005249079.2 | XP_005249136.1 | 13 | 1877 | |||
XM_006715082.3 | XP_006715145.1 | 12 | 3314 | |||
XM_011514567.2 | XP_011512869.1 | 13 | 2107 | |||
XM_017010819.1 | XP_016866308.1 | 13 | 1916 | |||
GenBank mRNA | AK122581.1 | - | 5 | 1023 | ||
AK127360.1 | - | 5 | 920 | |||
BC132831.1 | - | 12 | 1785 | 40146853 | ||
BC132833.1 | - | 12 | 1785 | 40146855 | ||
BX647582.1 | - | 13 | 2019 |