Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012975.1 | NP_001012993.1 | 1 | 223 | ||
GenBank mRNA | AK296766.1 | - | 4 | 711 | ||
AY665805.1 | - | 1 | 223 | |||
BC133002.1 | - | 1 | 242 | 40147024 | ||
BC133028.1 | - | 1 | 242 | 40147050 |